missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminotoluene 98.0+%, TCI America™

Catalog No. p-7128376
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46285G 5 g
D462825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46285G Supplier TCI America Supplier No. D46285G
Only null left

Chemical Identifiers

CAS 95-70-5
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00035779
InChI Key OBCSAIDCZQSFQH-UHFFFAOYSA-N
Synonym 2-Methyl-1,4-phenylenediamine
PubChem CID 7252
ChEBI CHEBI:53619
IUPAC Name 2-methylbenzene-1,4-diamine
SMILES CC1=C(C=CC(=C1)N)N

Specifications

Melting Point 62°C
Boiling Point 168°C
Quantity 5 g
UN Number 2811
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diaminotoluene
EINECSNumber (3)-0126
RTECSNumber XS9700000
TSCA Yes