missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminotoluene Sulfate 98.0+%, TCI America™

Catalog No. D160225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D160225G 25 g
D1602500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D160225G Supplier TCI America Supplier No. D160225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 615-50-9
Molecular Formula C7H12N2O4S
Molecular Weight (g/mol) 220.243
MDL Number MFCD00013003
InChI Key KZTWOUOZKZQDMN-UHFFFAOYSA-N
Synonym 2,5-diaminotoluene sulfate, 2-methylbenzene-1,4-diamine sulfate, fouramine std, toluene-2,5-diamine sulfate, 2,5-toluenediamine sulfate, 1,4-benzenediamine, 2-methyl-, sulfate, c.i. oxidation base 4, 2,5-diamino toluene sulfate, 2-methyl-p-phenylenediamine sulphate, 2-methyl-1,4-benzenediamine sulfate
PubChem CID 22856
IUPAC Name 2-methylbenzene-1,4-diamine;sulfuric acid
SMILES CC1=C(C=CC(=C1)N)N.OS(=O)(=O)O

Specifications

Color Red-White
Quantity 25 g
UN Number 2811
Formula Weight 220.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diaminotoluene Sulfate
RTECSNumber XT0525000
TSCA Yes