missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di(2-thienyl)thieno[3,2-b]thiophene 94.0+%, TCI America™

Catalog No. p-7128425
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4725200MG 200 mg
D47251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4725200MG Supplier TCI America Supplier No. D4725200MG
Only null left

Chemical Identifiers

CAS 21210-90-2
Molecular Formula C14H8S4
Molecular Weight (g/mol) 304.46
MDL Number MFCD11114556
InChI Key FDQXVHKNZBLBFY-UHFFFAOYSA-N
PubChem CID 11738270
IUPAC Name 2-[5-(thiophen-2-yl)thieno[3,2-b]thiophen-2-yl]thiophene
SMILES S1C=CC=C1C1=CC2=C(S1)C=C(S2)C1=CC=CS1

Specifications

Melting Point 254°C
Quantity 200 mg
Formula Weight 304.46
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di(2-thienyl)thieno[3,2-b]thiophene