missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile 95.0+%, TCI America™
$162.17 - $570.40
Chemical Identifiers
| CAS | 704890-84-6 |
|---|---|
| Molecular Formula | C12H5BrN2S2 |
| Molecular Weight (g/mol) | 321.21 |
| MDL Number | MFCD06214120 |
| InChI Key | GZLRCTASJSGPHL-UHFFFAOYSA-N |
| Synonym | 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]propanedinitrile |
| PubChem CID | 85975548 |
| IUPAC Name | 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile |
| SMILES | BrC1=CC=C(S1)C1=CC=C(S1)C=C(C#N)C#N |
Chemical Identifiers
| 704890-84-6 | |
| 321.21 | |
| GZLRCTASJSGPHL-UHFFFAOYSA-N | |
| 85975548 | |
| BrC1=CC=C(S1)C1=CC=C(S1)C=C(C#N)C#N |
| C12H5BrN2S2 | |
| MFCD06214120 | |
| 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]propanedinitrile | |
| 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile |
Specifications
| 704890-84-6 | |
| C12H5BrN2S2 | |
| 200 mg | |
| GZLRCTASJSGPHL-UHFFFAOYSA-N | |
| 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile | |
| 85975548 | |
| ≥95.0% (GC) | |
| 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]malononitrile |
| 202°C | |
| MFCD06214120 | |
| 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]propanedinitrile | |
| BrC1=CC=C(S1)C1=CC=C(S1)C=C(C#N)C#N | |
| 321.21 | |
| 321.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No