missing translation for 'onlineSavingsMsg'
Learn More

2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile 95.0+%, TCI America™

Catalog No. B5458200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5458200MG 200 mg
B54581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5458200MG Supplier TCI America Supplier No. B5458200MG
Only null left

Chemical Identifiers

CAS 704890-84-6
Molecular Formula C12H5BrN2S2
Molecular Weight (g/mol) 321.21
MDL Number MFCD06214120
InChI Key GZLRCTASJSGPHL-UHFFFAOYSA-N
Synonym 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]propanedinitrile
PubChem CID 85975548
IUPAC Name 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C=C(C#N)C#N

Specifications

Melting Point 202°C
Quantity 200 mg
Formula Weight 321.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(5′-Bromo-[2,2′-bithiophen]-5-yl)methylene]malononitrile