missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis[(trimethylsilyl)ethynyl]thiophene 96.0+%, TCI America™

Catalog No. B42405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42405G 5 g
B42401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42405G Supplier TCI America Supplier No. B42405G

Chemical Identifiers

CAS 79109-69-6
Molecular Formula C14H20SSi2
Molecular Weight (g/mol) 276.544
MDL Number MFCD18207392
InChI Key RAPGDWBCCYLGCE-UHFFFAOYSA-N
PubChem CID 12702007
IUPAC Name trimethyl-[2-[5-(2-trimethylsilylethynyl)thiophen-2-yl]ethynyl]silane
SMILES C[Si](C)(C)C#CC1=CC=C(S1)C#C[Si](C)(C)C

Specifications

Melting Point 82°C
Quantity 5 g
Formula Weight 276.54
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis[(trimethylsilyl)ethynyl]thiophene