missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene (purified by sublimation) 99.0+%, TCI America™

Catalog No. p-7123414
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42211G 1 g
B42215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42211G Supplier TCI America Supplier No. B42211G
Only null left

Chemical Identifiers

CAS 7128-64-5
Molecular Formula C26H26N2O2S
Molecular Weight (g/mol) 430.566
MDL Number MFCD00005774
InChI Key AIXZBGVLNVRQSS-UHFFFAOYSA-N
Synonym bbot, 2,5-bis 5-tert-butyl-2-benzoxazolyl thiophene, uvitex ob, 2,5-bis 5-tert-butyl benzo d oxazol-2-yl thiophene, bbot 150, 2,5-bis 5-tert-butylbenzoxazol-2-yl thiophene, benzoxazole, 2,2'-2,5-thiophenediyl bis 5-1,1-dimethylethyl, unii-w7ur9129gp, fluorescent brightener 184, 2,2'-2,5-thiophenediyl bis 5-tert-butylbenzoxazole
PubChem CID 292429
IUPAC Name 5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
SMILES CC(C)(C)C1=CC2=C(C=C1)OC(=N2)C3=CC=C(S3)C4=NC5=C(O4)C=CC(=C5)C(C)(C)C

Specifications

Melting Point 202°C
Quantity 1 g
Formula Weight 430.57
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene (purified by sublimation)
EINECSNumber (5)-3934
RTECSNumber DM4888332
TSCA Yes