missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole, 99%, Thermo Scientific Chemicals

Catalog No. p-7044706
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0787403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0787403 Supplier Thermo Scientific Chemicals Supplier No. L0787403

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2043-06-3
MDL Number MFCD00042666

Specifications

Melting Point 235°C to 238°C
Molecular Formula C26H18N2O
Quantity 1 g
Beilstein 318611
Synonym 2,5-bis 4-biphenylyl-1,3,4-oxadiazole, 2,5-bis 4-phenylphenyl-1,3,4-oxadiazole, 2,5-bis 1,1'-biphenyl-4-yl-1,3,4-oxadiazole, acmc-20aobr, 2,5-bis 4-biphenylyl oxadiazole, 2,5-bis biphenyl-4-yl-1,3,4-oxadiazole, 2,5-di-4-biphenylyl-1,3,4-oxadiazole, 2,5_di-4-biphenylyl-1,3,4-oxadiazole, 2,5-bis-4-biphenyl-yl-1,3,4-oxadiazole
InChI Key IVVYVRLKDGGUEL-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NN=C(O3)C4=CC=C(C=C4)C5=CC=CC=C5
IUPAC Name 2,5-bis(4-phenylphenyl)-1,3,4-oxadiazole
Molecular Weight (g/mol) 374.44
PubChem CID 96840
Formula Weight 374.45
Percent Purity 99%
Chemical Name or Material 2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole
Show More Show Less
EINECSNumber 218-046-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only