missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole 98.0+%, TCI America™

Supplier:  TCI America B21695G

 View more versions of this product

Catalog No. B21695G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole
Yellow
MFCD00042667
BAO
C1=CC(=CC=C1C2=NN=C(O2)C3=CC=C(C=C3)N)N
252.277
252.28
Crystalline Powder
2425-95-8
C14H12N4O
5 g
MJZXFMSIHMJQBW-UHFFFAOYSA-N
4-[5-(4-aminophenyl)-1,3,4-oxadiazol-2-yl]aniline
75505
≥98.0% (HPLC)

Chemical Identifiers

2425-95-8
252.277
MJZXFMSIHMJQBW-UHFFFAOYSA-N
75505
C1=CC(=CC=C1C2=NN=C(O2)C3=CC=C(C=C3)N)N
C14H12N4O
MFCD00042667
BAO
4-[5-(4-aminophenyl)-1,3,4-oxadiazol-2-yl]aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3589

TSCA : No

SDS
Documents

Documents

Promotions

Promotions