missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole 98.0+%, TCI America™

Catalog No. B21691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21691G 1 g
B21695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21691G Supplier TCI America Supplier No. B21691G

Chemical Identifiers

CAS 2425-95-8
Molecular Formula C14H12N4O
Molecular Weight (g/mol) 252.277
MDL Number MFCD00042667
InChI Key MJZXFMSIHMJQBW-UHFFFAOYSA-N
Synonym BAO
PubChem CID 75505
IUPAC Name 4-[5-(4-aminophenyl)-1,3,4-oxadiazol-2-yl]aniline
SMILES C1=CC(=CC=C1C2=NN=C(O2)C3=CC=C(C=C3)N)N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 252.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole
EINECSNumber (5)-3589
TSCA No