missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine 98.0+%, TCI America™
$201.32 - $613.27
Chemical Identifiers
| CAS | 152460-10-1 |
|---|---|
| Molecular Formula | C16H15N5 |
| Molecular Weight (g/mol) | 277.331 |
| MDL Number | MFCD09028125 |
| InChI Key | QGAIPGVQJVGBIA-UHFFFAOYSA-N |
| Synonym | N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine |
| PubChem CID | 9965429 |
| IUPAC Name | 4-methyl-3-N-(4-pyridin-3-ylpyrimidin-2-yl)benzene-1,3-diamine |
| SMILES | CC1=C(C=C(C=C1)N)NC2=NC=CC(=N2)C3=CN=CC=C3 |
Chemical Identifiers
| 152460-10-1 | |
| 277.331 | |
| QGAIPGVQJVGBIA-UHFFFAOYSA-N | |
| 9965429 | |
| CC1=C(C=C(C=C1)N)NC2=NC=CC(=N2)C3=CN=CC=C3 |
| C16H15N5 | |
| MFCD09028125 | |
| N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine | |
| 4-methyl-3-N-(4-pyridin-3-ylpyrimidin-2-yl)benzene-1,3-diamine |
Specifications
| 152460-10-1 | |
| Yellow | |
| MFCD09028125 | |
| N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine | |
| CC1=C(C=C(C=C1)N)NC2=NC=CC(=N2)C3=CN=CC=C3 | |
| 277.331 | |
| 277.33 | |
| Crystalline Powder |
| 145°C | |
| C16H15N5 | |
| 1 g | |
| QGAIPGVQJVGBIA-UHFFFAOYSA-N | |
| 4-methyl-3-N-(4-pyridin-3-ylpyrimidin-2-yl)benzene-1,3-diamine | |
| 9965429 | |
| ≥98.0% (HPLC,T) | |
| 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine |
Safety and Handling
TSCA : No