missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine 98.0+%, TCI America™

Catalog No. A23085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23085G 5 g
A23081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23085G Supplier TCI America Supplier No. A23085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 152460-10-1
Molecular Formula C16H15N5
Molecular Weight (g/mol) 277.331
MDL Number MFCD09028125
InChI Key QGAIPGVQJVGBIA-UHFFFAOYSA-N
Synonym N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine
PubChem CID 9965429
IUPAC Name 4-methyl-3-N-(4-pyridin-3-ylpyrimidin-2-yl)benzene-1,3-diamine
SMILES CC1=C(C=C(C=C1)N)NC2=NC=CC(=N2)C3=CN=CC=C3

Specifications

Melting Point 145°C
Color Yellow
Quantity 5 g
Formula Weight 277.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine