missing translation for 'onlineSavingsMsg'
Learn More

2,5,8,11,14-Pentaoxaheptadec-16-yne 98.0+%, TCI America™

Catalog No. P224925MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P224925MG 25 mg
P2249100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P224925MG Supplier TCI America Supplier No. P224925MG

Chemical Identifiers

CAS 1101668-39-6
Molecular Formula C12H22O5
Molecular Weight (g/mol) 246.30
MDL Number MFCD28155212
InChI Key QOXHTXZSGABRDF-UHFFFAOYSA-N
Synonym mPEG4-Alkyne
PubChem CID 86291528
IUPAC Name 2,5,8,11,14-pentaoxaheptadec-16-yne
SMILES COCCOCCOCCOCCOCC#C

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 246.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5,8,11,14-Pentaoxaheptadec-16-yne
TSCA No
Recommended Storage Refrigerator