missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Undecadien-1-ol (mixture of stereoisomers) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 168.28 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,4-Undecadien-1-ol (mixture of stereoisomers) |
Safety and Handling
| EINECSNumber | (2)-0258 |
| TSCA | Yes |