missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Thiazolyl)benzimidazole 98.0+%, TCI America™

Catalog No. T083025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T083025G 25 g
T0830250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T083025G Supplier TCI America Supplier No. T083025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148-79-8
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.247
MDL Number MFCD00005587
InChI Key WJCNZQLZVWNLKY-UHFFFAOYSA-N
Synonym thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole
PubChem CID 5430
ChEBI CHEBI:45979
IUPAC Name 4-(1H-benzimidazol-2-yl)-1,3-thiazole
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3

Specifications

Melting Point 305°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 201.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Thiazolyl)benzimidazole
EINECSNumber (9)-0820&(9)-1851
RTECSNumber DE0700000
TSCA Yes