missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Thiazolidinedione 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 128°C |
| Quantity | 100 g |
| Formula Weight | 117.12 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4-Thiazolidinedione |
Safety and Handling
| EINECSNumber | (5)-6527 |
| RTECSNumber | XJ5775000 |
| TSCA | Yes |