missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole 98.0+%, TCI America™
$162.17 - $162.17
Chemical Identifiers
| CAS | 15082-28-7 |
|---|---|
| Molecular Formula | C24H22N2O |
| Molecular Weight (g/mol) | 354.453 |
| MDL Number | MFCD00003101 |
| InChI Key | XZCJVWCMJYNSQO-UHFFFAOYSA-N |
| Synonym | butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole |
| PubChem CID | 84782 |
| IUPAC Name | 2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole |
| SMILES | CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4 |
Chemical Identifiers
| 15082-28-7 | |
| 354.453 | |
| XZCJVWCMJYNSQO-UHFFFAOYSA-N | |
| 84782 | |
| CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4 |
| C24H22N2O | |
| MFCD00003101 | |
| butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole | |
| 2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole |
Specifications
| 15082-28-7 | |
| White-Yellow | |
| MFCD00003101 | |
| butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole | |
| CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4 | |
| 354.453 | |
| 354.45 | |
| Crystalline Powder |
| 137°C | |
| C24H22N2O | |
| 5 g | |
| XZCJVWCMJYNSQO-UHFFFAOYSA-N | |
| 2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole | |
| 84782 | |
| ≥98.0% (HPLC) | |
| 2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole |
Safety and Handling
TSCA : Yes