missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pyridinedicarboxylic Acid 98.0+%, TCI America™

Catalog No. P24165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24165G 5 g
P241625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24165G Supplier TCI America Supplier No. P24165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 499-80-9 | C7H5NO4 | 167.12 g/mol

Chemical Identifiers

CAS 499-80-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00149413
InChI Key MJIVRKPEXXHNJT-UHFFFAOYSA-N
Synonym 2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
PubChem CID 10365
ChEBI CHEBI:44737
IUPAC Name pyridine-2,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC(=C1)C(O)=O

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Pyridinedicarboxylic Acid