missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Pentanedione Dioxime 98.0+%, TCI America™
Supplier: TCI America P079225G
Specifications
| 2,4-Pentanedione Dioxime | |
| White | |
| MFCD00013932 | |
| acetylacetone dioxime, 2,4-pentanedione dioxime, 2,4-pentanedione, dioxime, 2,4-pentanedioxime, 2,4-pentanedione oxime, 2,4-pentanedione,2,4-dioxime, 2,4-pentanedione, 2,4-dioxime, pentane-2,4-dione oxime, 2e,4e-2,4-pentanedione dioxime #, ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine | |
| C\C(C\C(C)=N\O)=N/O | |
| 130.15 | |
| 130.15 | |
| Crystalline Powder |
| 2157-56-4 | |
| C5H10N2O2 | |
| 25 g | |
| WBRYLZHYOFBTPD-YDFGWWAZSA-N | |
| (E)-N-[(4E)-4-(hydroxyimino)pentan-2-ylidene]hydroxylamine | |
| 9573501 | |
| ≥98.0% (N) |
Chemical Identifiers
| 2157-56-4 | |
| 130.15 | |
| WBRYLZHYOFBTPD-YDFGWWAZSA-N | |
| 9573501 | |
| C\C(C\C(C)=N\O)=N/O |
| C5H10N2O2 | |
| MFCD00013932 | |
| acetylacetone dioxime, 2,4-pentanedione dioxime, 2,4-pentanedione, dioxime, 2,4-pentanedioxime, 2,4-pentanedione oxime, 2,4-pentanedione,2,4-dioxime, 2,4-pentanedione, 2,4-dioxime, pentane-2,4-dione oxime, 2e,4e-2,4-pentanedione dioxime #, ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine | |
| (E)-N-[(4E)-4-(hydroxyimino)pentan-2-ylidene]hydroxylamine |
Safety and Handling
TSCA : Yes