missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Nitrophenyl)ethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. N064825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N064825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N064825G Supplier TCI America Supplier No. N064825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29968-78-3
Molecular Formula C8H11N2O2
Molecular Weight (g/mol) 167.19
MDL Number MFCD00012900
InChI Key IOXOZOPLBFXYLM-UHFFFAOYSA-O
Synonym 4-nitrophenethylamine hydrochloride, 4-nitrophenylethylamine hydrochloride, 4-nitrophenethylamine hcl, 2-4-nitrophenyl ethanamine hydrochloride, 2-4-nitrophenyl ethylamine hydrochloride, p-nitrophenylethylamine hcl, benzeneethanamine, 4-nitro-, monohydrochloride, 4-nitrophenethylaminehydrochloride, p-nitrophenethylamine, hydrochloride, pubchem11627
PubChem CID 458983
IUPAC Name 2-(4-nitrophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 214°C
Color Yellow
Quantity 25 g
Formula Weight 202.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Nitrophenyl)ethylamine Hydrochloride