missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Nitrophenyl)ethanol, 98%

Catalog No. p-7036243
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5450403 1 g
AAH5450406 5 g
AAH5450414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5450403 Supplier Thermo Scientific Chemicals Supplier No. H5450403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-27-6 | C8H9NO3 | 167.164 g/mol

It is a phosphate protecting reagent and O6 of deoxyguanosine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a phosphate protecting reagent and O6 of deoxyguanosine.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Keep at room temperature.

Chemical Identifiers

CAS 100-27-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00010202
InChI Key IKMXRUOZUUKSON-UHFFFAOYSA-N
Synonym 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
PubChem CID 7494
IUPAC Name 2-(4-nitrophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)[N+](=O)[O-]

Specifications

Melting Point 59°C to 62°C
Boiling Point 177°C (16 mmHg)
Quantity 1 g
Beilstein 1866148
Solubility Information Slightly soluble in water.
Formula Weight 167.16
Percent Purity 98%
Chemical Name or Material 2-(4-Nitrophenyl)ethanol
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 202-835-8
RTECSNumber SG8602000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only