missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Nitrophenyl)-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl Free Radical, TCI America™

Catalog No. D2295100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2295100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2295100MG Supplier TCI America Supplier No. D2295100MG
Only null left

Chemical Identifiers

CAS 38582-73-9
Molecular Formula C13H16N3O4
MDL Number MFCD00191637
Synonym 4,5-Dihydro-4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1H-imidazol-1-yloxy-3-oxide, 4-Nitrophenylnitronyl Nitroxide, p-NPNN

Specifications

Color Green
Quantity 100 mg
Formula Weight 278.29
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Nitrophenyl)-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl Free Radical