missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Morpholinyl)aniline, 98%

Catalog No. AAL0566006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0566006 5 g
AAL0566014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0566006 Supplier Thermo Scientific Chemicals Supplier No. L0566006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5585-33-1 | C10H14N2O | 178.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5585-33-1
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00047408
InChI Key QKWLVAYDAHQMLG-UHFFFAOYSA-N
Synonym 2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s
PubChem CID 735756
IUPAC Name 2-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=CC=C2N

Specifications

Melting Point 95°C to 98°C
Quantity 5 g
UN Number UN2811
Beilstein 163926
Formula Weight 178.24
Percent Purity 98%
Chemical Name or Material 2-(4-Morpholinyl)aniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only