missing translation for 'onlineSavingsMsg'
Learn More

{2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 879896-50-1
Molecular Formula C13H21N3
Molecular Weight (g/mol) 219.332
MDL Number MFCD06797802
InChI Key OKGWICVOLZFMPE-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
PubChem CID 16228064
IUPAC Name [2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine
SMILES CN1CCN(CC1)CC2=CC=CC=C2CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC47413CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC47413DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

879896-50-1
219.332
OKGWICVOLZFMPE-UHFFFAOYSA-N
16228064
CN1CCN(CC1)CC2=CC=CC=C2CN
C13H21N3
MFCD06797802
2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
[2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine

Specifications

879896-50-1
100.0
C13H21N3
2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
CN1CCN(CC1)CC2=CC=CC=C2CN
219.332
219.33
{2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine
96.5
97%
MFCD06797802
OKGWICVOLZFMPE-UHFFFAOYSA-N
[2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine
16228064
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents