missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC67504DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC67504DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

320423-50-5
211.283
TUJAFVJUJXMFEG-UHFFFAOYSA-N
1490234
CN1CCN(CC1)C2=NC=C(S2)C=O
C9H13N3OS
MFCD01568852
2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde

Specifications

2-(4-Methylpiperazin-1-yl)-1, 3-thiazole-5-carbaldehyde
96.5
97%
C9H13N3OS
MFCD01568852
TUJAFVJUJXMFEG-UHFFFAOYSA-N
2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
1490234
97%
320423-50-5
100.0
Amber Glass Bottle
1g
2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
CN1CCN(CC1)C2=NC=C(S2)C=O
211.283
211.29
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents