missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC67504CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67504CB 250mg Amber Glass Bottle
CC67504DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67504CB Supplier Thermo Scientific Chemicals Supplier No. CC67504CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 320423-50-5
Molecular Formula C9H13N3OS
Molecular Weight (g/mol) 211.283
MDL Number MFCD01568852
InChI Key TUJAFVJUJXMFEG-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
PubChem CID 1490234
IUPAC Name 2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
SMILES CN1CCN(CC1)C2=NC=C(S2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 211.29
Percent Purity 97%
Chemical Name or Material 2-(4-Methylpiperazin-1-yl)-1, 3-thiazole-5-carbaldehyde