missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)ethanol, 99%

Catalog No. AAB2091803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2091803 1 g
AAB2091806 5 g
AAB2091814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2091803 Supplier Thermo Scientific Chemicals Supplier No. B2091803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 699-02-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 699-02-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002904
InChI Key DAVFJRVIVZOKKS-UHFFFAOYSA-N
Synonym 2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol
PubChem CID 95294
IUPAC Name 2-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)CCO

Specifications

Density 0.98
Boiling Point 244°C to 245°C
Flash Point 107°C (225°F)
Refractive Index 1.526
Quantity 1 g
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 2-(4-Methylphenyl)ethanol
EINECSNumber 211-824-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only