missing translation for 'onlineSavingsMsg'
Learn More

[2-(4-Methylperhydro-1,4-diazepin-1-yl)phenyl]methanol, ≥97%, Thermo Scientific™

Catalog No. CC60409DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC60409DA 1g Amber Glass Bottle
CC60409DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC60409DA Supplier Thermo Scientific Chemicals Supplier No. CC60409DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 915707-55-0
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
MDL Number MFCD09065000
InChI Key IYBDLMFCIKDQDV-UHFFFAOYSA-N
Synonym 2-4-methylperhydro-1,4-diazepin-1-yl phenyl methanol, 2-4-methyl-1,4-diazepan-1-yl phenyl methanol, 2-4-methylhomopiperazin-1-yl benzyl alcohol, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methan-1-ol
PubChem CID 24229618
IUPAC Name [2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanol
SMILES CN1CCCN(CC1)C1=CC=CC=C1CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material [2-(4-Methylperhydro-1, 4-diazepin-1-yl)phenyl]methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.