missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-5-thiazolyl)ethyl Decanoate 98.0+%, TCI America™

Catalog No. M24905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24905G 5 g
M249025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24905G Supplier TCI America Supplier No. M24905G

Chemical Identifiers

CAS 101426-31-7
Molecular Formula C16H27NO2S
Molecular Weight (g/mol) 297.46
MDL Number MFCD09032915
InChI Key RUOLKIITSCGKJQ-UHFFFAOYSA-N
Synonym Decanoic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Decanoyloxyethyl)-4-methylthiazole
PubChem CID 44182035
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl decanoate
SMILES CCCCCCCCCC(=O)OCCC1=C(C)N=CS1

Specifications

Color Yellow
Boiling Point 181°C
Quantity 5 g
Formula Weight 297.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl Decanoate