missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%

Catalog No. p-7026075
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2186706 5 g
AAB2186714 25 g
AAB2186722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2186706 Supplier Thermo Scientific Chemicals Supplier No. B2186706

CAS: 656-53-1 | C8H11NO2S | 185.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 656-53-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.24
MDL Number MFCD00005338
InChI Key CRTCWNPLKVVXIX-UHFFFAOYSA-N
Synonym 2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole
PubChem CID 61192
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate
SMILES CC(=O)OCCC1=C(C)N=CS1

Specifications

Density 1.147
Boiling Point 116°C to 118°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.51
Quantity 5 g
Formula Weight 185.24
Percent Purity ≥98%
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl acetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-515-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only