missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-1-piperazinyl)aniline 98.0+%, TCI America™

Catalog No. p-7133311
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25921G 1 g
M25925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25921G Supplier TCI America Supplier No. M25921G
Only null left

Chemical Identifiers

CAS 180605-36-1
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.278
MDL Number MFCD04035359
InChI Key INWHDRNGZMHXEZ-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl aniline, 2-4-methyl-1-piperazinyl aniline, 2-4-methylpiperazino aniline, 2-4-methyl-piperazin-1-yl-phenylamine, benzenamine, 2-4-methyl-1-piperazinyl, 1-2-aminophenyl-4-methylpiperazine, 2-4-methylpiperazinyl phenylamine, asischem z51464, pubchem13000, akos bb-7895
PubChem CID 286547
IUPAC Name 2-(4-methylpiperazin-1-yl)aniline
SMILES CN1CCN(CC1)C2=CC=CC=C2N

Specifications

Melting Point 105°C
Color Yellow
Quantity 1 g
UN Number 3263
Formula Weight 191.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Methyl-1-piperazinyl)aniline