missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methoxyphenyl)pyridine 97.0+%, TCI America™
$78.29 - $870.51
Chemical Identifiers
| CAS | 5957-90-4 |
|---|---|
| Molecular Formula | C12H11NO |
| Molecular Weight (g/mol) | 185.226 |
| MDL Number | MFCD00092333 |
| InChI Key | QUMXRZNAUFKBAS-UHFFFAOYSA-N |
| Synonym | 4-(2-Pyridyl)anisole |
| PubChem CID | 220239 |
| IUPAC Name | 2-(4-methoxyphenyl)pyridine |
| SMILES | COC1=CC=C(C=C1)C2=CC=CC=N2 |
Chemical Identifiers
| 5957-90-4 | |
| 185.226 | |
| QUMXRZNAUFKBAS-UHFFFAOYSA-N | |
| 220239 | |
| COC1=CC=C(C=C1)C2=CC=CC=N2 |
| C12H11NO | |
| MFCD00092333 | |
| 4-(2-Pyridyl)anisole | |
| 2-(4-methoxyphenyl)pyridine |
Specifications
| 5957-90-4 | |
| White-Yellow | |
| C12H11NO | |
| 1 g | |
| QUMXRZNAUFKBAS-UHFFFAOYSA-N | |
| 2-(4-methoxyphenyl)pyridine | |
| 220239 | |
| ≥97.0% (GC,T) | |
| 2-(4-Methoxyphenyl)pyridine |
| 55°C | |
| 130°C | |
| MFCD00092333 | |
| 4-(2-Pyridyl)anisole | |
| COC1=CC=C(C=C1)C2=CC=CC=N2 | |
| 185.226 | |
| 185.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No