missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7132365
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M079525ML 25 mL
M0795250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M079525ML Supplier TCI America Supplier No. M079525ML
Only null left

Chemical Identifiers

CAS 55-81-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00008192
InChI Key LTPVSOCPYWDIFU-UHFFFAOYSA-N
Synonym 4-(2-Aminoethyl)anisole, 4-Methoxyphenethylamine
PubChem CID 4657
ChEBI CHEBI:266039
IUPAC Name 2-(4-methoxyphenyl)ethanamine
SMILES COC1=CC=C(C=C1)CCN

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 mL
UN Number 2735
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4-Methoxyphenyl)ethylamine
RTECSNumber SH7875000
TSCA Yes