missing translation for 'onlineSavingsMsg'
Learn More

2-(4-methoxyphenoxy)ethanamine hydrochloride, 97%, Thermo Scientific™

Catalog No. BTB10107DA
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB10107DA 1g
BTB10107EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB10107DA Supplier Thermo Scientific Chemicals Supplier No. BTB10107DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98959-77-4
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.666
InChI Key YCFXTJDWVNEVEX-UHFFFAOYSA-N
Synonym 2-4-methoxyphenoxy ethan-1-amine hydrochloride, 2-4-methoxyphenoxy ethanamine hydrochloride, 2-4-methoxyphenoxy ethylamine hydrochloride, 2-4-methoxyphenoxy ethylaminehydrochloride, 1-2-aminoethoxy-4-methoxybenzene hydrochloride
PubChem CID 43810691
IUPAC Name 2-(4-methoxyphenoxy)ethanamine;hydrochloride
SMILES COC1=CC=C(C=C1)OCCN.Cl

Specifications

Assay Percent Range 97%
Quantity 1g
Formula Weight 203.67
Chemical Name or Material 2-(4-methoxyphenoxy)ethanamine hydrochloride