missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxybenzoyl)thiophene 98.0+%, TCI America™

Catalog No. M23955G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23955G Supplier TCI America Supplier No. M23955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 4160-63-8
Melting Point 74°C
Molecular Formula C12H10O2S
Quantity 5 g
MDL Number MFCD00005440
Synonym 2-4-methoxybenzoyl thiophene, 4-methoxyphenyl thiophen-2-yl methanone, 2-p-methoxybenzoyl thiophene, p-anisyl thiophen-2-yl ketone, 4-methoxyphenyl 2-thienyl ketone, methanone, 4-methoxyphenyl-2-thienyl, 4-methoxyphenyl 2-thienyl methanone, pubchem10833, acmc-1aikh, 2-thienyl p-anisyl ketone
InChI Key KYVBFEMQEUXVQB-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CS2
IUPAC Name (4-methoxyphenyl)-thiophen-2-ylmethanone
Molecular Weight (g/mol) 218.27
PubChem CID 77812
Formula Weight 218.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Methoxybenzoyl)thiophene
Show More Show Less