missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxybenzoyl)thiophene, 97%

Catalog No. AAL0551318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0551318 50 g
AAL0551309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0551318 Supplier Thermo Scientific Chemicals Supplier No. L0551318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4160-63-8 | C12H10O2S | 218.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4160-63-8
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00005440
InChI Key KYVBFEMQEUXVQB-UHFFFAOYSA-N
Synonym 2-4-methoxybenzoyl thiophene, 4-methoxyphenyl thiophen-2-yl methanone, 2-p-methoxybenzoyl thiophene, p-anisyl thiophen-2-yl ketone, 4-methoxyphenyl 2-thienyl ketone, methanone, 4-methoxyphenyl-2-thienyl, 4-methoxyphenyl 2-thienyl methanone, pubchem10833, acmc-1aikh, 2-thienyl p-anisyl ketone
PubChem CID 77812
IUPAC Name (4-methoxyphenyl)-thiophen-2-ylmethanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CS2

Specifications

Melting Point 71°C to 75°C
Quantity 50 g
Beilstein 148560
Formula Weight 218.27
Percent Purity 97%
Chemical Name or Material 2-(4-Methoxybenzoyl)thiophene
EINECSNumber 223-997-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only