missing translation for 'onlineSavingsMsg'
Learn More

2,4-Lutidine, 98+%, Thermo Scientific™

Chemical Identifiers

CAS 108-47-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006337
InChI Key JYYNAJVZFGKDEQ-UHFFFAOYSA-N
Synonym 2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
PubChem CID 7936
IUPAC Name 2,4-dimethylpyridine
SMILES CC1=CC=NC(C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125181000
SDS View Documents
Thermo Scientific Chemicals
125181000
100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC125185000
SDS View Documents
Thermo Scientific Chemicals
125185000
500mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

108-47-4
107.16
JYYNAJVZFGKDEQ-UHFFFAOYSA-N
7936
CC1=CC=NC(C)=C1
C7H9N
MFCD00006337
2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
2,4-dimethylpyridine

Specifications

108-47-4
100.0
159.0°C
98% min. (GC)
C7H9N
MFCD00006337
, 244
0.92
Solubility in water: 15g/100 g (20°C). Other solubilities: soluble in alcohol,ether,acetone and,benzene
CC1=CC=NC(C)=C1
107.16
107.15
2, 4-Lutidine, 98+%
98.0
0.9200g/mL
47°C
Glass bottle
1.4990 to 1.5010
100mL
06,224
2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
JYYNAJVZFGKDEQ-UHFFFAOYSA-N
2,4-dimethylpyridine
7936
98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Danger

EINECSNumber : 203-586-8

RTECSNumber : OK9400000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only