missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isocyanatophenyl)-5-(trifluoromethyl)pyridine, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC53006DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC53006DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

906352-73-6
264.207
GFEVPIXAPFROGH-UHFFFAOYSA-N
24229533
C1=CC(=CC=C1C2=NC=C(C=C2)C(F)(F)F)N=C=O
C13H7F3N2O
MFCD09817476
2-4-isocyanatophenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl-2-pyridyl benzenisocyanate
2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine

Specifications

2-(4-Isocyanatophenyl)-5-(trifluoromethyl)pyridine
96.5
97+%
C13H7F3N2O
MFCD09817476
GFEVPIXAPFROGH-UHFFFAOYSA-N
2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine
24229533
97%
906352-73-6
100.0
Amber Glass Bottle
1g
2-4-isocyanatophenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl-2-pyridyl benzenisocyanate
C1=CC(=CC=C1C2=NC=C(C=C2)C(F)(F)F)N=C=O
264.207
264.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents