missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isocyanatophenyl)-5-(trifluoromethyl)pyridine, ≥97%, Thermo Scientific™

Catalog No. CC53006CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53006CB 250mg Amber Glass Bottle
CC53006DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC53006CB Supplier Thermo Scientific Chemicals Supplier No. CC53006CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 906352-73-6
Molecular Formula C13H7F3N2O
Molecular Weight (g/mol) 264.207
MDL Number MFCD09817476
InChI Key GFEVPIXAPFROGH-UHFFFAOYSA-N
Synonym 2-4-isocyanatophenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl-2-pyridyl benzenisocyanate
PubChem CID 24229533
IUPAC Name 2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine
SMILES C1=CC(=CC=C1C2=NC=C(C=C2)C(F)(F)F)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 264.21
Percent Purity 97%
Chemical Name or Material 2-(4-Isocyanatophenyl)-5-(trifluoromethyl)pyridine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.