missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isobutylphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. I041525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I041525G 25 g
I0415100G 100 g
I0415500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I041525G Supplier TCI America Supplier No. I041525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
Synonym ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
PubChem CID 3672
ChEBI CHEBI:5855
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

Melting Point 76°C
Color White
Boiling Point 157°C
Quantity 25 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Isobutylphenyl)propionic Acid
EINECSNumber (9)-0183
RTECSNumber MU6640000
TSCA Yes