missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Isobutylphenyl)propionic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 76°C |
| Color | White |
| Boiling Point | 157°C |
| Quantity | 100 g |
| Formula Weight | 206.29 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(4-Isobutylphenyl)propionic Acid |
Safety and Handling
| EINECSNumber | (9)-0183 |
| RTECSNumber | MU6640000 |
| TSCA | Yes |