Learn More
2-(4-Isobutylphenyl)propanenitrile, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 58609-73-7 |
|---|---|
| Molecular Formula | C13H17N |
| Molecular Weight (g/mol) | 187.286 |
| MDL Number | MFCD02180706 |
| InChI Key | PKQKHWNHCKNYSW-UHFFFAOYSA-N |
| Synonym | 2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrile |
| PubChem CID | 108374 |
| ChEBI | CHEBI:27415 |
| IUPAC Name | 2-[4-(2-methylpropyl)phenyl]propanenitrile |
| SMILES | CC(C)CC1=CC=C(C=C1)C(C)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
SEW06499DA
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 58609-73-7 | |
| 187.286 | |
| PKQKHWNHCKNYSW-UHFFFAOYSA-N | |
| 108374 | |
| 2-[4-(2-methylpropyl)phenyl]propanenitrile |
| C13H17N | |
| MFCD02180706 | |
| 2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrile | |
| CHEBI:27415 | |
| CC(C)CC1=CC=C(C=C1)C(C)C#N |
Specifications
| 58609-73-7 | |
| 97% | |
| C13H17N | |
| MFCD02180706 | |
| PKQKHWNHCKNYSW-UHFFFAOYSA-N | |
| 2-[4-(2-methylpropyl)phenyl]propanenitrile | |
| 108374 | |
| 187.28 | |
| 2-(4-Isobutylphenyl)propanenitrile |
| 97.0 | |
| Amber Glass Bottle | |
| (CH3)2CHCH2C6H4CH(CN)CH3 | |
| 2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrile | |
| CC(C)CC1=CC=C(C=C1)C(C)C#N | |
| 187.286 | |
| CHEBI:27415 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.