missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)pyridine 98.0+%, TCI America™

Catalog No. p-7129859
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07271G 1 g
F07275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07271G Supplier TCI America Supplier No. F07271G
Only null left

Chemical Identifiers

CAS 58861-53-3
Molecular Formula C11H8FN
Molecular Weight (g/mol) 173.19
MDL Number MFCD06201382
InChI Key MHWIDTQQBWGUCD-UHFFFAOYSA-N
PubChem CID 100868
IUPAC Name 2-(4-fluorophenyl)pyridine
SMILES C1=CC=NC(=C1)C2=CC=C(C=C2)F

Specifications

Melting Point 40°C
Boiling Point 119°C
Quantity 1 g
Formula Weight 173.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Fluorophenyl)pyridine