missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)indole 98.0+%, TCI America™

Catalog No. F04275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04275G Supplier TCI America Supplier No. F04275G

Specifications

CAS 782-17-2
Melting Point 190°C
Color Yellow
Molecular Formula C14H10FN
Quantity 5 g
MDL Number MFCD00068178
Synonym 2-4-fluorophenyl indole, 2-4-fluorophenyl-1h-indole, pubchem7339, acmc-209pdr, maybridge1_007619, 2-4-fluoro-phenyl-1h-indole
InChI Key VLHGDCJIDNVRFM-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=C(N2)C3=CC=C(C=C3)F
IUPAC Name 2-(4-fluorophenyl)-1H-indole
Molecular Weight (g/mol) 211.239
PubChem CID 136622
Formula Weight 211.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Fluorophenyl)indole
Show More Show Less