missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. F08295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08295G 5 g
F082925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08295G Supplier TCI America Supplier No. F08295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1583-88-6
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD00134208
InChI Key CKLFJWXRWIQYOC-UHFFFAOYSA-N
Synonym 4-fluorophenethylamine, 2-4-fluorophenyl ethanamine, 2-4-fluoro-phenyl-ethylamine, p-fluorophenethylamine, 4-fluorophenylethylamine, 2-4-fluorophenyl ethylamine, 2-4-fluorophenyl ethan-1-amine, benzeneethanamine, 4-fluoro, p-fluorophenyl ethylamine, 2-4-fluorophenyl-ethylamine
PubChem CID 4653
IUPAC Name 2-(4-fluorophenyl)ethanamine
SMILES C1=CC(=CC=C1CCN)F

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 2735
Formula Weight 139.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4-Fluorophenyl)ethylamine