missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)ethylamine, 97%

Catalog No. AAH6179122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6179122 100 g
AAH6179114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6179122 Supplier Thermo Scientific Chemicals Supplier No. H6179122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1583-88-6 | C8H10FN | 139.173 g/mol

2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 1583-88-6
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD00134208
InChI Key CKLFJWXRWIQYOC-UHFFFAOYSA-N
Synonym 4-fluorophenethylamine, 2-4-fluorophenyl ethanamine, 2-4-fluoro-phenyl-ethylamine, p-fluorophenethylamine, 4-fluorophenylethylamine, 2-4-fluorophenyl ethylamine, 2-4-fluorophenyl ethan-1-amine, benzeneethanamine, 4-fluoro, p-fluorophenyl ethylamine, 2-4-fluorophenyl-ethylamine
PubChem CID 4653
IUPAC Name 2-(4-fluorophenyl)ethanamine
SMILES C1=CC(=CC=C1CCN)F

Specifications

Color Yellow
Density 1.061 g/mL
Boiling Point 50°C to 52°C
Flash Point 79°C (174°F)
Refractive Index 1.507
Quantity 100 g
UN Number UN2922
Solubility Information Slightly soluble in water.
Formula Weight 139.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(4-Fluorophenyl)ethylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 216-435-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only