missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)ethyl Chloride 97.0+%, TCI America™

Catalog No. F04781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04781G Supplier TCI America Supplier No. F04781G
Only null left

Chemical Identifiers

CAS 332-43-4
Molecular Formula C8H8ClF
Molecular Weight (g/mol) 158.60
MDL Number MFCD00155000
InChI Key LPUURRVXQCVXCX-UHFFFAOYSA-N
Synonym 1-2-chloroethyl-4-fluorobenzene, 4-fluorophenethyl chloride, 2-4-fluorophenyl ethyl chloride, 1-2'-chloroethyl-4-fluorobenzene, 1-2-chloro-ethyl-4-fluoro-benzene, benzene, 1-2-chloroethyl-4-fluoro, acmc-1anql, 4-fluorophenetyl chloride, ksc570e7d, 1-fluoro-4-2-chloroethyl benzene
PubChem CID 67622
IUPAC Name 1-(2-chloroethyl)-4-fluorobenzene
SMILES FC1=CC=C(CCCl)C=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 158.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Fluorophenyl)ethyl Chloride