missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7130045
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1045200MG 200 mg
F10451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1045200MG Supplier TCI America Supplier No. F1045200MG

Chemical Identifiers

CAS 203450-08-2
Molecular Formula C21H14FN3
Molecular Weight (g/mol) 327.36
MDL Number MFCD28964710
InChI Key DEFTZOFCLNDIMR-UHFFFAOYSA-N
PubChem CID 15850059
IUPAC Name 2-(4-fluorophenyl)-4,6-diphenyl-1,3,5-triazine
SMILES FC1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 233°C
Color White
Quantity 200 mg
Formula Weight 327.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine