missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenoxy)nicotinic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 54629-13-9
Molecular Formula C12H8FNO3
Molecular Weight (g/mol) 233.198
MDL Number MFCD00833409
InChI Key SDZUYDOXBXHDCE-UHFFFAOYSA-N
Synonym 2-4-fluorophenoxy nicotinic acid, 2-4-fluorophenoxy pyridine-3-carboxylic acid, 2-4-fluoro-phenoxy-nicotinic acid, pubchem16292, acmc-1akqk, maybridge1_000048, 2-4-fluorophenoxy-3-pyridinecarboxylic acid, 3-pyridinecarboxylicacid, 2-4-fluorophenoxy, 3-pyridinecarboxylic acid, 2-4-fluorophenoxy
PubChem CID 605123
IUPAC Name 2-(4-fluorophenoxy)pyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
TL00878EA
SDS View Documents
Thermo Scientific Chemicals
TL00878EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

54629-13-9
233.198
SDZUYDOXBXHDCE-UHFFFAOYSA-N
605123
C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O
C12H8FNO3
MFCD00833409
2-4-fluorophenoxy nicotinic acid, 2-4-fluorophenoxy pyridine-3-carboxylic acid, 2-4-fluoro-phenoxy-nicotinic acid, pubchem16292, acmc-1akqk, maybridge1_000048, 2-4-fluorophenoxy-3-pyridinecarboxylic acid, 3-pyridinecarboxylicacid, 2-4-fluorophenoxy, 3-pyridinecarboxylic acid, 2-4-fluorophenoxy
2-(4-fluorophenoxy)pyridine-3-carboxylic acid

Specifications

54629-13-9
97%
C12H8FNO3
10g
SDZUYDOXBXHDCE-UHFFFAOYSA-N
2-(4-fluorophenoxy)pyridine-3-carboxylic acid
605123
97%
97.0
Amber Glass Bottle
MFCD00833409
2-4-fluorophenoxy nicotinic acid, 2-4-fluorophenoxy pyridine-3-carboxylic acid, 2-4-fluoro-phenoxy-nicotinic acid, pubchem16292, acmc-1akqk, maybridge1_000048, 2-4-fluorophenoxy-3-pyridinecarboxylic acid, 3-pyridinecarboxylicacid, 2-4-fluorophenoxy, 3-pyridinecarboxylic acid, 2-4-fluorophenoxy
C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O
233.198
233.2
2-(4-Fluorophenoxy)nicotinic acid
Videos
SDS
Documents

Documents