missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%

Catalog No. AAH5441006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5441006 5 g
AAH5441003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5441006 Supplier Thermo Scientific Chemicals Supplier No. H5441006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28485-18-9 | C4H3ClN2O4S | 210.59 g/mol

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 28485-18-9
Molecular Formula C4H3ClN2O4S
Molecular Weight (g/mol) 210.59
MDL Number MFCD06655485
InChI Key FQHNJCWWBKYCJW-UHFFFAOYSA-N
Synonym 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride, 1,2,3,4-tetrahydro-2,4-dioxo-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonylchloride, 1,2,3,4-tetrahydro-2,4-dioxo, uracil-5-sulphonyl chloride, 5-chlorosulfonyl-1,3-dihydropyrimidine-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinesulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydro-pyrimidine-5-sulfonyl chloride
PubChem CID 234620
SMILES ClS(=O)(=O)C1=CNC(=O)NC1=O

Specifications

Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
Formula Weight 209.95
Percent Purity 97%
Chemical Name or Material 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only